ULTRA-WARE® Filtering Flask, Kimble Chase
Filter Flask with Host Connection, 1 L, 8, 2
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 953760-0000
ULTRA-WARE® Filtering Flask, Kimble Chase
Catalog # KT953760-0000
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
This flask is manufactured from heavy wall borosilicate glass to assure the mechanical strength needed for vacuum filtration
The side arm accepts standard 1/4" (6 mm) I.D. tubing for connection to vacuum sources. A second flask should be connected between the filtering flask and the vacuum source to prevent accidental entry of the filtrate into the vacuum line or pump.
Especificaciones
- Capacity:1 L
- Description:Filter Flask with Host Connection
- Stopper No.:8
- Connection Size:2