Orders LinkContactUsLinkComponent
Microscope Slide Storage Boxes for 100 Slides, Globe Scientific
Microscope Slide Storage Boxes for 100 Slides, Globe Scientific
Microscope Slide Storage Boxes for 100 Slides, Globe Scientific
Certificates
Microscope Slide Storage Boxes for 100 Slides, Globe Scientific
Microscope Slide Storage Boxes for 100 Slides, Globe Scientific
3 Options Available Below
Product Details & Documents
Globe's microscope slide storage boxes are designed for the safe and long term storage of microscope slides
An inventory sheet is located on the inside cover for cenvenient slide identification and organization. The base of the box is cork lined for added protection.
They are produced from durable ABS plastic and offer a hinged lid for easy opening and closing.
Ordering information: Available in 4 sizes, these storage boxes accommodate 25 x 75 mm and 3 x 1" microscope slides.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Dark Grey
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $213.48
    $213.48
    Each (1UN)
       $213.48
    Supplier Number: 513079G
  • Grey
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $213.48
    $213.48
    Each (1UN)
       $213.48
    Supplier Number: 513079A
  • Orange
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $213.48
    $213.48
    Each (1UN)
       $213.48
    Supplier Number: 513079N