SP Bel-Art 72-Piece Complete Fitting Assortment, Bel-Art Products, a part of SP
470123-372
- Descripción:72-Piece Fitting Kit
- Referencia:470123-372
- Nº de proveedor:F19625-0000
This generous assortment of 72 plastic fittings has everything you need for routine laboratory set-ups.
- Clear Polystyrene Construction
- Hinged Lid
- Translucent Siding
- Seventy-Two Plastic Fittings Included
- Autoclavable Fittings
A clear, hinge-top, polystyrene storage box makes it easy to find what you need when you need it.
The kit includes common sizes of connector and stopper styles.
: Polypropylene fittings are FDA-graded for direct contact with food, drugs, and cosmetics and are autoclavable.
: This assortment includes polypropylene connectors: 5 pairs of straight tubing connectors and 4 pairs of T connectors. It has three polyethylene connectors: 3 pairs of quick disconnects, 6 pairs of straight-stepped tubing connectors, and 6 pairs of y connectors. It has 12 pairs of cup-type, polyethylene stoppers.