Whatman™ Kjeldahl Analysis Weighing Paper, Whatman products (Cytiva)
Proveedor: Cytiva
Certificados
Acerca de este artículo
Grade B-2 Whatman weighing paper from Cytiva dissolves residue-free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
- Thin (0.04 mm) and lightweight (43 g/m²)
- Transparent and smooth parchment surface
- Simple weighing out and sample transfer
- Reliable for quantitative transfer
- Range of sheet sizes for ease-of-use
- Low nitrogen paper that is residue-free for accurate chemical analysis
Cytiva also offers weighing boats for Kjeldahl analysis.
Packs of 500 Grade B-2 Whatman weighing paper are available in four sheet sizes: 76.2×76.2 mm (3×3 inch) sheets; 101.6×101.6 mm (4×4 inch) sheets; 152.4×152.4 mm (6×6 inch) sheets; 304.8×304.8 mm (12×12 inch) sheets.
Información relacionada
Certificados
Buscar por número de loteCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together