96-Well PCR Tube Racks
71003-652
- Ø orificio:5.2 mm
- Núm. de orificios:96
- Color:Assorted
- Descripción:96-well PCR racks
- Dimensiones:130×98×33 mm (5×3.9×1.2")
- Referencia:71003-652
- For Tubes:0.2 ml
- Nº de proveedor:HS23461A
Perfect for pre- or post-PCR sampling.
- PCR rack is made of polypropylene
- Accommodates individual tubes or strips
- Removable hinged lid lies flat
- Alphanumeric identification for quick location and retrieval of tubes
These sturdy polypropylene racks accommodate individual 0.2 ml tubes.
: Assorted color orders include blue, green, orange, purple, and yellow racks.