Corning® Polystyrene Storage Bottles, Corning
500 ml (16.9 oz.), Polystyrene Storage Bottles with Male MPC Connectors, Sterile
Supplier: Corning
Número de proveedor #: 11666
Corning® Polystyrene Storage Bottles, Corning
Catalog # 89501-310
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Corning polystyrene storage bottles are now available as closed system solutions.
- Sterile
- Arrive individually double-bagged and peassembled
The preassembled transfer caps have two ports. One port ends in a 0.2 µm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in a Male Luer Lock. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
Especificaciones
- Sterility:Sterile
- Capacity:500 ml (16.9 oz.)
- Description:Polystyrene Storage Bottles with Male MPC Connectors
- Cat. No.:89501-310
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact