Simple Machines, Screw Model
Simple Machines, Screw Model
Catalog # 470210-826
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: WDMS20
Jump to:
Product Details & Documents
This wooden machine has a sturdy wooden construction ideal for classroom use.
- Demonstrate the concept of leverage
- Includes manual
The large block mounted around the head of the screw gives students additional leverage so they can easily break eggs or squish other objects with the screw that would be difficult to break with their bare hands. The square top also makes it easy to measure the distance top of the screw is turned vs. the distance the screw is sent into the board. Includes an instruction manual to facilitate students and teacher use of the apparatus.
Especificaciones
- Description:Simple Machines, Screw Model