Orders LinkContactUsLinkComponent
Corning® Pasteur Pipettes
Corning® Pasteur Pipettes
Supplier:   Corning
Corning® Pasteur Pipettes
Supplier:  Corning
Certificates
Corning® Pasteur Pipettes
Corning® Pasteur Pipettes
1 Options Available Below
Product Details & Documents
Soda-lime or borosilicate glass.
  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures.
Product Family Options
Información del producto
  • Material
    Ext.Ø
    Length
    Sterility
    Availability
    Price
  • 14673-156
    Disposable pasteur pipettes
    Disposable pasteur pipettes
    Soda-lime glass
    6.5 mm
    146.05 mm (5¾")
    Non sterile
    Discontinued
    Supplier Number: -
  • 89090-042
    Disposable pasteur pipettes
    Disposable pasteur pipettes
    Borosilicate glass
    6.5 mm
    146.05 mm (5¾")
    Non sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $3,600.00
    $3,600.00
    Case (1,000UN)
       $3,600.00
    Supplier Number: 7095D-5X
  • 52947-802
    Disposable pasteur pipettes
    Disposable pasteur pipettes
    Soda-lime glass
    5.5 mm
    228.6 mm (9")
    Non sterile
    Discontinued
    Supplier Number: -