Eisco® Chicken Models
Rooster, 1, 18 cm, 28 cm, 34 cm
Catalog # 470136-418
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: ZM0026
Jump to:
Product Details & Documents
The naturally sized, three-part, cross-sectional chicken models depict the bird in high detail.
- Cross-Sectional
- Naturally Sized
- Removable Internal Organs and Muscular System
Students can remove the internal organs and muscular system.
It's easy to closely examine both the internal and external anatomical structures.
Documents
470212-486_key
Documentación del producto
Especificaciones
- Description:Rooster
- Height:34 cm
- Length:28 cm
- Width:18 cm
- No. of Parts:1