SP Bel-Art Write-On™ Bottles, HDPE, Bel-Art Products, a part of SP
Catalog # 74950-033
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacidad:500 ml (16.9 oz.)
- Descripción:Wide Mouth Write-On™ Bottles
- Referencia:74950-033
- Nº de proveedor:106620500
Specifications
About this item
Molded from rugged high-density polyethylene in 500 and 1000 ml (16.9 and 33.8 oz.) sizes, these bottles are available in narrow (28 mm) or wide mouth (53 mm) formats, and have leakproof polypropylene caps.
- Rugged HDPE construction
- Leakproof polypropylene caps
- Ideal for use with both liquids or dry materials
Screened labels include a name identification area and a large detail area for contents or safety information, as well as a defined space for date information. Ideal for storing and transporting liquids or dry material.