Especificaciones
- Descripción:Memory-Card Refrigerator/Freezer Thermometer
- Precisión de la temperatura:±0.5°C from 0°C to 25°C, ±0.6°C from 25°C to 50°C, ±1.2°C from -30°C to 0°C and 50°C to 70°C
- Temperature resolution:0.1°
- Humidity range:0 to 99.9% RH
- Precisión de la humedad:±5% RH at 25°C for range of 20 to 85% RH, ±7% RH elsewhere
- Referencia:89369-154
- Sonda:Bottle probe
- Rango:–30…+70° C (–22…+158 °F)
Especificaciones
Acerca de este artículo
Traceable® Memory-Card Refrigerator/Freezer Thermometer is a stand-alone unit with data logging capabilities. It records readings from once a minute up to once every 720 minutes on a removable standard SD memory card. Thermometers captures 5.9 million readings with the supplied 256 mb removable SD memory card. Memory card reader plugs into any USB computer port for an instant report. Text report includes date or time-of-day and sensor readings, it may be sent to any spreadsheet or database. Capturing a continuous stream of data shows the behavior of refrigerators, room areas or tests. The bottle probe model is ideal for monitoring refrigerators, freezers and incubators; the stainless steel probe model is ideal for QC and research. Bottle probe meets all CDC, federal and state refrigerator thermometer requirements. Both models include internal sensors to measure the ambient temperature and humidity - ideal for monitoring store rooms, labs and critical areas.
- Insulated from rapid temperature changes
- Does not disturb seal of closed doors
- Unique 4-line display
- Captures a continuous stream of data
- Records up to 5.9 million readings
It records readings from once a minute up to once every 720 minutes on a removable standard SD memory card
Sealed bottle sensor is insulated from rapid temperature changes when refrigerator door is opened. Solution in sealed bottle is generally recognized as safe by the FDA. The cable is easy for the refrigerator door to close and seal. Minimum/Maximum feature displays highest/lowest readings for any time period. Alarm sound and flashes a bright red LED when temperature rises above or falls below user-set trip points, alarm sets is 0.1° increments. Unique 4-line display shows temperature of probe, ambient temperature, humidity and time of day.
Thermometers captures 5.9 million readings with the supplied 256 mb removable SD memory card. Memory card reader plugs into any USB computer port for an instant report. Text report includes date or time-of-day and sensor readings, it may be sent to any spreadsheet or database. Capturing a continuous stream of data shows the behavior of refrigerators, room areas or tests. The bottle probe model is ideal for monitoring refrigerators, freezers and incubators; the stainless steel probe model is ideal for QC and research. Bottle probe meets all CDC, federal and state refrigerator thermometer requirements. Both models include internal sensors to measure the ambient temperature and humidity - ideal for monitoring store rooms, labs and critical areas.