ContactUsLinkComponent
Filter Adapters, Neoprene, Chemglass
Filter Adapters, Neoprene, Chemglass
Catalog # 80061-064
Supplier:  CHEMGLASS INC.
Small Business Enterprise
undefined
Filter Adapters, Neoprene, Chemglass
Catalog # 80061-064
Supplier:  CHEMGLASS INC.
Supplier Number:  CG-1401-13
Small Business Enterprise
Specifications
  • Descripción:
    Filter Adapter, Size 4, 39 x 22 mm I.D., 46 x 29 mm O.D., 3.5 mm thickness
  • Referencia:
    80061-064
  • Nº de proveedor:
    CG-1401-13
Specifications
About this item
Neoprene filter adapters are designed to fit between the filtering funnel and flask.
For use with Buchner funnels, Grooch crucibles, filter tubes, etc. Can be used individually or nesting with adjacent sizes. Eliminates the boring of rubber stoppers and the difficulties of inserting and removing glass stems in stoppers. Neoprene adapters withstand autoclaving.