Micro Capillary Tubes, Disposable, Kimble Chase
10 µl, Orange
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Número de proveedor #: 71900-10
Micro Capillary Tubes, Disposable, Kimble Chase
Catalog # 72690-002
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Capillary action draws the fluid into the tube. To dispense, simply cover the hole on the rubber bulb and squeeze. Pipettes are 5" long with a single black capacity ring. They are color-coded for selection of the correct size.
- Packaged in easy-open cylinders for convenient dispensing
- Clean and ready to use
- Filling is easy
Certifications: Micro Capillary pipettes made of soda lime glass in compliance with ASTM Specification E438, Type II glass requirements.
Packaging: Packaging is designed from ASTM Specification E672 requirements. Each cylinder contains 250 pipettes and one pipette device.
Documents
Especificaciones
- Capacity:10 µl
- Color:Orange