VWR® Cleanroom Sponges
A range of sponges, made with cleanroom materials, are designed for cleaning small parts, equipment, and other items in production or the lab.
- Durable, versatile, and absorbent range of sponges
- Hold up to alcohols, common cleaning solutions, and strong disinfectants
- Reusable and autoclavable
Fabric cover options include a 100% polyester braided mesh, a cleanroom alternative to abrasive pads to remove residues, a 100% polyester knit, FDA approved for indirect food contact, and a white microfiber 80% polyester/20% polyamide split fiber fabric with high absorbency and superior cleaning ability.
Frequently Bought Together