SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Catalog # 70680-270
Supplier: Bel-Art Products, a Part of SP
Specifications
- Tamaño:76 cm (30") wide x 15.2 meter (50 ft)
- Referencia:70680-270
- Nº de proveedor:246790000
Specifications
About this item
Clear polyethylene ribbed matting can be used for lining laboratory tables, shelves, trays, etc.
- Clear polyethylene construction
- Ribbed design
- Easily trimmed to fit laboratory surfaces
It is easily trimmed with scissors and also works well under drums or carboys. 0.25 mm (0.010") with 0.7 mm (0.030") thick ribs.