ContactUsLinkComponent
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Catalog # 89497-822
undefined
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Catalog # 89497-822
Supplier Number:  110444
Specifications
  • Ø×L:
    12×75 mm
  • Volumen:
    5 ml
  • Descripción:
    Test Tube, Conical Bottom
  • Matriz:
    PP
  • Referencia:
    89497-822
  • Nº de proveedor:
    110444
Specifications
About this item
These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.
Ordering information: Unless otherwise specified, test tubes are supplied without caps.