Self Standing Sample Tubes, 5 and 10 ml, Globe Scientific
Catalog # 89497-742
Supplier: GLOBE SCIENTIFIC INC
Specifications
- Ø×L:16×92 mm
- Volumen:10 ml
- Descripción:Transport Tube with Separate White Screw Cap
- Referencia:89497-742
- Estilo:Conical Bottom, Self Standing
- Nº de proveedor:6102
Specifications
About this item
Transport tubes are designed for the safe storage and transport of biological materials.
- Tubes are manufactured from polypropylene and feature a skirted conical bottom base and molded graduations
- Caps are manufactured from polyethylene
These transparent polypropylene vials allow unobstructed viewing of the sample. They are shatter-proof and may be exposed to freezing temperatures.
The leak-resistant screw caps can be removed or sealed in a single turn.