Orders LinkContactUsLinkComponent
Cryogenic Vials, Internal or External Threaded, Simport Scientific
Cryogenic Vials, Internal or External Threaded, Simport Scientific
Certificates
Cryogenic Vials, Internal or External Threaded, Simport Scientific
Cryogenic Vials, Internal or External Threaded, Simport Scientific
1 Options Available Below
Product Details & Documents
Designed for storing biological material, human or animal cells at temperatures as low as -196℃. Should be used only in the gas phase of liquid nitrogen. Available in 2 different configurations and in 6 sizes from 1.2 mL to 10 mL. Each has a large white marking area and printed graduations. Closures and tubes are both made of polypropylene having the same coefficient of expansion which further enhances the leakproof qualities of these vials at various temperatures. Please note that model T310-10A has a polyethylene screw cap.
  • Large white marking area and printed graduations
  • Model T310-10A has a polyethylene screw cap
Closures and tubes are both made of polypropylene having the same coefficient of expansion which further enhances the leakproof qualities of these vials at various temperatures.
 : Certified Rnase-, DNase-, Pyrogen- and DNA-free. CE marked.
 : Tubes can be color coded by the use of CAPINSERT Accessory.
 : Packaged in tamperproof, resealable, safety-lock bags of 100.
 : These vials should be used only in the gas phase of liquid nitrogen.
Product Family Options
Internal Threaded
Información del producto
  • Capacity
    Bottom Style
    Diameter ×H
    Cap included
    Sterility
    Availability
    Price
  • 10062-820
    With silicone o-ring seal cap
    With silicone o-ring seal cap
    4.00 ml
    Round bottom
    12.5×70 mm
    Yes
    Sterile
    Discontinued
    Supplier Number: T301-4
  • 73960-418
    With silicone washer seal cap
    With silicone washer seal cap
    4.00 ml
    Round bottom
    12.5×70 mm
    Yes
    Sterile
    Log in Para disponibilidad
    Log in Para disponibilidad
    Case (1,000UN)
    Case (1,000UN) - $10,047.07
    $10,047.07
    Case (1,000UN)
       $10,047.07
    Supplier Number: T311-4