ContactUsLinkComponent
Accessories for Socorex™ Acura® electro Variable Volume Pipettes, DWK Life Sciences
Accessories for Socorex™ Acura® electro Variable Volume Pipettes, DWK Life Sciences
Catalog # 53498-666
Small Business Enterprise
undefined
Accessories for Socorex™ Acura® electro Variable Volume Pipettes, DWK Life Sciences
Catalog # 53498-666
Supplier Number:  851346
Small Business Enterprise
Specifications
  • Descripción:
    Nozzle protection filters for 10 ml model, polypropylene fibers
  • Referencia:
    53498-666
  • Para ser utilizado con:
    Nozzle filters for Acura® electro 926 XS 10 ml pipette
  • Nº de proveedor:
    851346