Tite-Rite™ Containers, Globe Scientific
90 ml (3 oz.), 53 mm, PP
10030-358
: 6525
Tite-Rite™ containers provide the ultimate convenience and reliability in leak-tight storage and liquid transport. The specially designed caps contain an inner liner designed to compress against the uppermost walls of the container, ensuring an air-tight seal. The containers are ideal for use in hospital pneumatic tube systems.
Manufactured from polypropylene, these conatiners are autoclavable at 121°C (250°F) for 20 minutes (without caps). Containers are 95kPa compliant. Caps are manufactured from polyethylene.
The specially designed caps contain an inner liner designed to compress against the uppermost walls of the container, ensuring an air-tight seal. The containers are ideal for use in hospital pneumatic tube systems.
- Cap Size:53 mm
- Capacity:90 ml (3 oz.)
- Material:PP