TouchNTuff® 93-700, Sterile Nitrile Gloves, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Certificates
About this item
TouchNTuff® Sterile Nitrile Gloves feature an enhanced nitrile formulation for chemical protection, flexibility, comfort, and dexterity.
- Ambidextrous
- Excellent chemical protection
- Low allergenicity
Free of natural rubber latex proteins.
Suggested applications include core pharmaceutical products manufacturing, biotechnology, medical devices, academic research instituitions, and R&D laboratories.
Gamma radiation – sterility assurance level (Sal 10⁻⁶). The 93-700 is an ambidextrous glove with a roll-beaded cuff. Clean sterile pair packs with a pinhole AQL of 1.0. Optimal for critical environment applications.
Certifications: Manufactured in a facility registered to ISO 9001, ISO 13485, ISO14001. Quality control complies with ASTM D 3577, Type II.
Specifications
- Catalog No:
- 89402-742
- 89402-740
- 89402-744
- Ambidiestro/Específico para cada mano:
- Ambidiestro
- Ambidiestro
- Ambidiestro
- Desechable / Reutilizable:
- Desechable
- Desechable
- Desechable
- Acabado:
- Textured
- Textured
- Textured