Inlet Adapter, Glassco
89426-420
- Descripción:Inlet Adapter
- Referencia:89426-420
- ST Joint Size:24/40
- Nº de proveedor:036.472.03
Inlet adapters permit the insertion of plain stem thermometers or bleed tubes.
- Accommodates thermometer or bleed tube
- Equipped with one-holed rubber adapter
Inlet adapter features a standard taper joint.