Orders LinkContactUsLinkComponent
PLC Plates, MilliporeSigma
PLC Plates, MilliporeSigma
PLC Plates, MilliporeSigma
PLC plates allow users to separate samples that vary greatly in size - from grams down to milligrams
Product # 1.05434.0001 replaces and is identical to Product # 5434-7.
Product # 1.05637.0001 replaces and is identical to Product # 5637-7.
Product # 1.05717.0001 replaces and is identical to Product # 5717-7.
Product # 1.05726.0001 replaces and is identical to Product # 5726-7.
Product # 1.05744.0001 replaces and is identical to Product # 5744-7.
Product # 1.05745.0001 replaces and is identical to Product # 5745-7.
Product # 1.13894.0001 replaces and is identical to Product # 13894-7.
Product # 1.13895.0001 replaces and is identical to Product # 13895-7.
Available with or without a fluorescence indicator. They also use the same proven silica-binder technology as in analytical TLC plates. To isolate the substance by extraction, users can simply scrape the spot from the layer.
Product Family Options
  • Coating material
    Plate size
    Layer thickness
    Pore size
    Fluorescent Indicator
    No. of Plates
  • Silica gel
    200×200 mm
    2000 µm
    60 Å
    No
    12
    Each (1UN)
    Each (1UN) - $17,438.05
    $17,438.05
    Each (1UN)
       $17,438.05
     : 1057450001
  • Silica gel
    200×200 mm
    500 µm
    60 Å
    F254
    20
    Each (1UN)
    Each (1UN) - $13,430.90
    $13,430.90
    Each (1UN)
       $13,430.90
     : 1057440001
  • Silica gel
    200×200 mm
    1000 µm
    60 Å
    F254
    15
    Each (1UN)
    Each (1UN) - $19,389.69
    $19,389.69
    Each (1UN)
       $19,389.69
     : 1138950001