Filter Funnel, Buchner, with Inner Joint, Glassco
89426-762
- Porosidad:Medium
- [ST] unión No.:24/40
- Capacidad:60 ml
- Descripción:Filter Funnel, Buchner, with Inner Joint
- Referencia:89426-762
- Nº de proveedor:257.M00.02AB
Buchner filter funnel with a lower inner joint and serrated tubulation for connection to a vacuum source
Not recommended for filtration where pressure exceeds 15psi.