Filtering Flask, Microscale, Glassco
Filtering Flask, Microscale, 25 ml
Catalog # 89426-772
Supplier: GLASSCO LABORATORY EQUIPMENT
Número de proveedor #: 259.471.01
Jump to:
Product Details & Documents
Flask top is tooled for a #2 neoprene filter adapter or #3 rubber stopper
Manufactured from Borosilicate 3.3 glass.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:25 ml
- Description:Filtering Flask, Microscale