Round Bottom Flask, Angled, Triple Neck, Glassco
Catalog # 89426-546
Supplier: GLASSCO LABORATORY EQUIPMENT
Specifications
- [ST] unión No.:24/40
- Capacidad:500 ml
- Descripción:Round Bottom Flask, Angled, Triple Neck
- Referencia:89426-546
- Side Neck ST Joints:24/40
- Nº de proveedor:060.470.09
Specifications
About this item
Round bottom flask with joints
Heavy wall design. Manufactured from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform wall thickness.