Orders LinkContactUsLinkComponent
Shaking Water Baths, Circulating/Reciprocating, Shel Lab Sheldon
Shaking Water Baths, Circulating/Reciprocating, Shel Lab Sheldon
Water Bath, Circulating, Reciprocating/Shaking, 27 L, 115V
Catalog # 89409-392
Small Business Enterprise
Número de proveedor #:  SWBR27
Shaking Water Baths, Circulating/Reciprocating, Shel Lab Sheldon
Catalog # 89409-392
Small Business Enterprise
Shaking Water Baths, Circulating/Reciprocating, Shel Lab Sheldon
Shaking Water Baths, Circulating/Reciprocating, Shel Lab Sheldon
Water Bath, Circulating, Reciprocating/Shaking, 27 L, 115V
Price
$146,136.03
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
SHEL LAB Reciprocating Water Baths deliver precise temperature control and a smooth reciprocal shaking motion
The bath may be used with or without oscillation. Shaking speed is variable from 20 to 200 oscillations per minute, and the stroke length can be adjusted to three separate settings: 0.5, 1.0, or 1.5 inches to minimize the chance of accidental spillage. Also, the unique basket support system is designed to eliminate tank wear, yet still provide full support and free action for a heavily loaded basket.
A durable finish on the housing prevents corrosion and chipping. In addition to a stainless steel tank, the bath platform tray is designed for optimum temperature uniformity. Maximum temperature is achieved using the stainless steel gable cover, which is designed to prevent condensate from dropping onto samples and assures uniform temperature conditions.
Temperature and oscillation controls feature an easy to read P.I.D. microprocessor temperature controller and an independent safety thermostat.
Ordering information: 2 Year Limited Warranty (excludes tank). One stainless steel gable included.
Documents
Especificaciones
  • Description:
    Water Bath, Circulating, Reciprocating/Shaking, 27 L, 115V
  • Cat. No.:
    89409-392
  • Temperature uniformity:
    +/- 0.2 °C at 37 °C
  • Shaking speed:
    20 - 200 min⁻¹ (oscillation rate)
  • Int. W×D×H:
    68.5 x 30.4 x 18.4 cm (27 x 12 x 7.2"), Bath
  • Platform:
    62.2 x 27.9 x 19.6 cm (24.5 x 11 x 7.7")
  • Capacity:
    27 L (Tank)
  • Model:
    27 L
  • Temperature range:
    5 °C above Ambient to 80 °C
  • Electrical:
    115V
  • Ext. W×D×H:
    95.3 x 37.5 x 31.8 cm (37.5 x 14.8 x 12.5")