ContactUsLinkComponent
VACUUBRAND® Oil-Free Vacuum Pumps for Filtration and Solid Phase Extraction, BrandTech
VACUUBRAND® Oil-Free Vacuum Pumps for Filtration and Solid Phase Extraction, BrandTech
Catalog # 97058-166
undefined
VACUUBRAND® Oil-Free Vacuum Pumps for Filtration and Solid Phase Extraction, BrandTech
Catalog # 97058-166
Supplier Number:  20721103
Specifications
  • Descripción:
    VACUUBRAND® ME1C Vacuum Pump, 100-120V, 50/60 Hz, US plug
  • Referencia:
    97058-166
  • Ultimate vacuum:
    75 torr (100 mbar)
  • Free Air Capacity:
    14 L/min (0.5 CFM)
  • Nº de proveedor:
    721103
Specifications
About this item
The new ME1 and ME1C diaphragm vacuum pumps offer a compact, high performance and easy-to-use solution which is perfect for both single and multiple filtrations, solid phase extraction (SPE) and other fluid movement tasks
  • Excellent for filtration, degassing and SPE
  • Quiet, low vibration
  • Choose ME 1C for acidic vapors
  • Typical service intervals >15,000 operating hours
  • Optional flow valve/manometer
Corrosion resistant design. Both models feature robust PTFE diaphragms and valves for excellent corrosion resistance in routine biology and chemistry lab applications. The ME1 has an aluminum head and is well suited for aqueous and some organic solvents. The ME1C incorporates a PTFE head to provide a full fluoropolymer flow path for maximum chemical resistance to the most challenging applications, including acidic vapors.
Quiet and reliable; these pumps offer whisper-quiet operation, low vibration, and long service intervals; typically 15,000 operating hours in most applications.
Accessories; matching vacuum regulator “bleeder” valves with integral Bourdon vacuum gauges are available for both models.
The functional, space saving and innovative design with readily accessible top mounted power switch ensures convenient and quick operation.
Certifications: cTUVus.
Ordering information: Pumps are supplied with 100-120V, 50/60Hz motors, US plug, Operating Manual and One Year Warranty.