Peroxide Test Method: Colorimetric With Test Strips 100 - 200 - 400 - 600 - 800 - 1000 mg/L H₂O₂ MQuant™, MilliporeSigma
Catalog # EM1.10337.0001
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Método químico:Enzymatic reaction
- Descripción:Test with Strips
- Referencia:EM1.10337.0001
- Rango:100-200-400-600-800-1000 mg/l H₂O₂
- Nº de proveedor:1.10337.0001
- No. of Tests:100 strips
Specifications
About this item
The peroxide test is suitable for checking the current concentration of diluted peroxide solutions to ensure the disinfectant effect.
MilliporeSigma # 1.10337.0001 replaces and is identical to MilliporeSigma # 10337-1