Reflectoquant® Hydroxymethylfurfural (HMF) Test, MilliporeSigma
Catalog # EM1.17952.0001
Supplier: MERCK, S.A. DE C.V. SV
CAS Number:
Specifications
- Descripción:Reflectoquant® Hydroxymethylfurfural (HMF) Test Method: Reflectometric with Test Strips
- Referencia:EM1.17952.0001
- Rango:1.0 - 60.0 mg/L
- Nº de proveedor:1.17952.0001
- No. de tiras:50
Specifications
About this item
Method: Reflectometric With Test Strips 1.0 - 60.0 mg/l Reflectoquant® HMF Test.
Reflectoquant® Hydroxymethylfurfural (HMF) is the first easy-to-use rapid test of HMF, an important quality parameter in thermally treated fruit juices and honey samples. Enzymatic reaction.
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.