C-MAG HP 10 IKATHERM® Hot Plate, IKA® Works
Catálogo # 89495-344
Proveedor: IKA WORKS, INC. MX
cUL Listed
UL Listed
Especificaciones
- Rango de temperatura:50–500°C (122–932°F)
- Descripción:C-MAG HP 10 IKATHERM® Hot Plate
- Dimensiones:300W x 105D x 415H mm (113/4 x 41/8 x 163/8")
- Peso:6 kg (13.2 lbs.)
- Referencia:89495-344
- Detalles eléctricos:115 V, 50/60 Hz
- Material de la placa superior:Ceramic
- Nº de proveedor:3582001
Especificaciones
Acerca de este artículo
Newly designed hotplate constructed of ceramic glass to offer excellent chemical resistance.
Standard features include a fixed safety circuit of 550 °C, Hot Top indicator for hot surface warning to prevent burns, exact temperature setting via digital display (LED), a digital error code display, and an elevated control panel for protection against liquid leaks. Also provided is a bushing according to DIN 12878 for connecting a contact thermometer (see 89028-114 series) to enable precise temperature control.
Units meet IP 21 protection class according to DIN EN 60529.
Certificaciones: CE marked. UL and C-UL listed.
Información de pedido: Supplied with a three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). For additional laboratory accesory options, see the IKA® Lablift Manual (see 89495-302 series). Accessories are sold separately.