C-MAG HP 7 IKATHERM® Hot Plate, IKA® Works
89495-340
cUL Listed
UL Listed
- Rango de temperatura:50–500°C (122–932°F)
- Descripción:C-MAG HP 7 IKATHERM® Hot Plate
- Dimensiones:220W x 105D x 330H mm (85/8 x 41/8 x 13")
- Peso:5 kg (11 lbs.)
- Referencia:89495-340
- Detalles eléctricos:115 V, 50/60 Hz
- Material de la placa superior:Ceramic
- Nº de proveedor:3581801
Newly designed hotplate constructed of ceramic glass to offer excellent chemical resistance.
Standard features include a fixed safety circuit of 550 °C, Hot Top indicator for hot surface warning to prevent burns, exact temperature setting via digital display (LED), a digital error code display, and an elevated control panel for protection against liquid leaks. Also provided is a bushing according to DIN 12878 for connecting a contact thermometer (see 89028-114 series) to enable precise temperature control.
Units meet IP 21 protection class according to DIN EN 60529.
: CE marked. UL and C-UL listed.
: Supplied with a three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). For additional laboratory accessory options, see the IKA® Lablift Manual (see 89495-302 series). Accessories are sold separately.