Orders LinkContactUsLinkComponent
Disposable Polyethylene Dropper Pipettes
Disposable Polyethylene Dropper Pipettes
Disposable Polyethylene Dropper Pipettes
Certificates
Disposable Polyethylene Dropper Pipettes
Disposable Polyethylene Dropper Pipettes
4 Options Available Below
Product Details & Documents
These polyethylene pipettes are chemically inert.
  • For consistent bulb pressure and top circumference
  • Disposable
Ensure precisely measured drop reproducibility.
Product Family Options
Información del producto
  • Capacity
    Length
    Stem Diameter
    Bulb Draw
    Availability
    Price
  • 470016-028
    Dropper pipette, graduated, 6573305
    Dropper pipette, graduated, 6573305
    1.5 ml
    114 mm
    1.5 mm
    0.8 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 500
    Paquete de 500 - $700.30
    $700.30
    Log in Para disponibilidad
    Case de 10 (500UN)
    Case de 10 (500UN) - $763.33
    $763.33
    Paquete de 500
       $700.30
    Case de 10 (500UN)
       $763.33
    Supplier Number: P200-14
  • 470105-218
    Dropper pipette, graduated, 6573301
    Dropper pipette, graduated, 6573301
    1.5 ml
    114 mm
    1.5 mm
    0.8 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $63.00
    $63.00
    Paquete de 100
       $63.00
    Supplier Number: -
  • 470144-238
    Dropper pipette, micro tip, 6525405
    Dropper pipette, micro tip, 6525405
    5 ml
    114 mm
    5 mm
    3.3 ml
    Discontinued
    Supplier Number: -
  • 470190-802
    Dropper pipette, micro tip, 6525410
    Dropper pipette, micro tip, 6525410
    5 ml
    114 mm
    5 mm
    3.3 ml
    Discontinued
    Supplier Number: -
  • 470190-800
    Dropper pipette, thin stem, 6572800
    Dropper pipette, thin stem, 6572800
    4.5 ml
    150 mm
    2.5 mm
    3.3 ml
    Discontinued
    Supplier Number: -
  • 470175-296
    Dropper pipette, micro tip/jumbo bulb, 6509100
    Dropper pipette, micro tip/jumbo bulb, 6509100
    8.9 ml
    150 mm
    3 mm
    4.8 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 400
    Paquete de 400 - $1,067.39
    $1,067.39
    Log in Para disponibilidad
    Case de 10 (400UN)
    Case de 10 (400UN) - $1,163.46
    $1,163.46
    Paquete de 400
       $1,067.39
    Case de 10 (400UN)
       $1,163.46
    Supplier Number: 234
  • 470144-498
    Dropper pipette, extra long, 6509000
    Dropper pipette, extra long, 6509000
    6 ml
    225 mm
    5 mm
    2.3 ml
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 500
    Paquete de 500 - $798.12
    $798.12
    Log in Para disponibilidad
    Case de 10 (500UN)
    Case de 10 (500UN) - $869.95
    $869.95
    Paquete de 500
       $798.12
    Case de 10 (500UN)
       $869.95
    Supplier Number: P200-60
  • Discontinued
    Supplier Number: -