PIG® Mercury Spill Vacuum Kit, New Pig
Catalog # 56620-340
Supplier: 2 D 2 S.A. DE C.V. MX
Specifications
- Capacidad:Up to 177.4 ml (6 oz.)
- Referencia:56620-340
- Nº de proveedor:KIT330
Specifications
About this item
Vacuum pump allows you to quickly and effectively capture mercury spills for proper recycling or disposal
- Scoop and spatula make it easier to gather scattered mercury droplets without unnecessary scattering or loss
- Durable polyethylene carrying case keeps all necessary equipment in one place
- No hazardous components to generate additional hazardous waste
No wheels. Not UN rated.
Includes collection vials and cleanup gear for up to 6 spills