PIG® HazMat Spill Kit in High-Visibility Bag, New Pig
Catalog # 56620-494
Supplier: 2 D 2 S.A. DE C.V. MX
Specifications
- Capacidad:Up to 34.1 L (9 gal.)
- Referencia:56620-494
- Nº de proveedor:KIT320
Specifications
About this item
Durable, water-resistant bag with hook-and-loop closure keeps contents clean and dry
Temporary disposal bags help make cleanup easier. Bright yellow bag and large, color-coded label make the kit easy to locate. Strap for easy carrying and hanging. Compact and easy to store; makes a great truck spill kit.
PIG HazMat Socks stop spreading spills; PIG HazMat Pads absorb quickly. PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide