SeQuant® ZIC®-HILIC HPLC Columns, MilliporeSigma
Catálogo # 10144-132
Proveedor: MERCK, S.A. DE C.V. SV
Especificaciones
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:3.5 µm
- Tamaño de poro:100 Å
- Descripción:ZIC®-HILIC HPLC column
- Longitud:150 mm
- Column Hardware:Acero inoxidable, revestimiento de PEEK
- Referencia:10144-132
- Packing Material:Silica
- Phase:HILIC
- Nº de proveedor:1.50444.0001
Especificaciones
Acerca de este artículo
The selectivity offered by ZIC®-HILIC (Hydrophilic Interaction Liquid Chromatography) is suitable for a wide variety of molecules containing hydrophilic or ionisable functional groups.
- Straightforward separation of polar and hydrophilic compounds
- Orthogonal selectivity compared to reversed-phase
- High-density bonded sulfobetaine zwitterionic stationary phase
- Low cost per analysis due to robust stationary phase
This includes compounds such as carbohydrates, metabolites, acids and bases, organic and inorganic ions, metal complexes, amino acids, peptides, protein digests, plant and cell extracts, plus much more. These compounds are normally characterised by a small or negative LogP (Octanol-water partition coefficient) value and have poor retention on reversed-phase columns.