Spill-X-C® Caustic-Neutralizing Adsorbent, New Pig
89205-860
- Descripción:Pail, 50 lbs.
- Referencia:89205-860
- Nº de proveedor:GEN323
Neutralizes and solidifies many caustics for safe and easy cleanup, including sodium hydroxide (50%) and ammonium hydroxide (29%)
Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime. Excellent for use in shipping/receiving areas, caustic container storage areas and caustic use and transfer sites. Non-toxic, free-flowing dry chemical is environmentally friendly.