PIG® Poly Modular Spill Deck, New Pig
Unique "flow-through" sump design lets liquid pass freely from one sump to another to help you comply with containment regulations. Modular design features exclusive bulkhead fittings and predrilled ports to connect to other decks; ready to use as soon as they arrive. Mix and match the sections you need to fit your space and help meet sump capacity requirements. Special tools and hardware to connect sections is included with each system.
- Unique "flow-through" sump lets liquid pass freely from one sump to another to comply with containment regulations
- Bulkhead fittings and predrilled ports connect to other decks
- Mix and match sections to fit your space and help meet sump capacity requirements
- Special tools and hardware to connect sections included with each system
- Low-density polyethylene (LDPE) resists UV rays, rust, corrosion and most chemicals
- Textured grating adds traction and lifts out for easy sump access
- Translucent white sidewalls allow easy visual inspection
Low-profile 5.75"H design is easy to load or unload; pumps and funnels are easy to access
Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals. Textured grating adds traction and lifts out for easy sump access. Translucent white sidewalls allow easy visual inspection.
Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe.
- Description:Holds two 55-gal. steel or poly drum
- Load Capacity:3000 lbs.
- Sump Capacity:22 gal.