ContactUsLinkComponent
SP Bel-Art Lab Companion™ Cabinet Style Vacuum Desiccators, Bel-Art Products, a part of SP
SP Bel-Art Lab Companion™ Cabinet Style Vacuum Desiccators, Bel-Art Products, a part of SP
Catalog # 30621-068
undefined
SP Bel-Art Lab Companion™ Cabinet Style Vacuum Desiccators, Bel-Art Products, a part of SP
Catalog # 30621-068
Supplier Number:  F42400-4031
Specifications
  • Volumen:
    45 L (1.6 cu. ft.), internal
  • Color:
    Clear
  • Referencia:
    30621-068
  • Dimensiones exteriores:
    42.0W × 39.7D × 49.1H cm (16½ × 15⅝ × 19⅜")
  • No. de bandejas:
    3 incl., 6 max.
  • Door Opening:
    29.5W × 44.5H cm (11⅝ × 17½")
  • Peso:
    12.2 kg (26.75 lbs.)
  • Nº de proveedor:
    F42400-4031
  • Shelf Capacity:
    5 kg (11 lbs.) max.
Specifications
About this item
These high-quality vacuum desiccators stay gastight allowing reliable dry storage or experiments over extended periods.
  • Hinged door design
  • Movable perforated shelves
  • Transparent polycarbonate construction
  • Amber-tined models protect light-sensitive samples
Able to maintain 29in.-Hg vacuum for over 72 hours at room temperature, they provide vacuum integrity resulting in lower maintenance and greater peace of mind
Constructed of transparent polycarbonate, these desiccators are durable, easy-to-clean, chemical-resistant, and allow quick visualization of the desiccator contents. Amber-tinted UV-blocking models block 100% of UVA, B, and C light to minimize damages or discoloration of light-sensitive samples.
The hinged door allows easy access and built in locking tabs secure the door when not under vacuum. Perforated shelves can be removed or repositioned to accommodate various storage needs (additional shelves sold separately). Built-in ports permit attachment of up to 3 additional valves (sold separately). A desiccant tray is included (desiccants sold separately).
Single piece molded design (except the door) enables outstanding vacuum capability with virtually no leakage and exceptional durability while the cabinet style provides maximum useable space and is stackable. A Viton® 3-way valve offers consistent and uniform vacuum draw, vacuum release, or gas exchange. An analog vacuum gauge in front door allows vacuum level checking at a glance.
Ordering information: Use with any vacuum with free air capacity of 1.5 cubic feet per minute (CFM) ft or greater. Eight models to choose from; 11, 23, 35, and 45 liter internal volume in clear or UV-blocking amber.