Sabdex Agar with Chloramphenicol, CRITERION™, Hardy Diagnostics
Catálogo # 89405-976
Proveedor: HARDY DIAGNOSTICS
Product available on GSA Advantage®
Número CAS:
Especificaciones
- Tipo de env.:Bottle, wide mouth
- Tamaño:500 g
- Temperatura de almacenamiento:–2-30°C
- Referencia:89405-976
- Nº de proveedor:C6781
Especificaciones
Acerca de este artículo
CRITERION™ Sabdex Agar with Chloramphenicol is recommended as a selective agar medium for the isolation, identification and cultivation of fungi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certificaciones: BSE Free