SP Bel-Art Stack Rack™ Test Tube Racks, Bel-Art Products, a part of SP
60917-462
- Núm. de orificios:24 (3×8)
- Descripción:Polypropylene Stack Rack™ Test Tube Rack
- Dimensiones:29.7L x 12.7W x 13.3H cm (11¹¹/₁₆ x 5 x 5¹/₄")
- Referencia:60917-462
- Para D.E. de tubo:26 - 30 mm
- Nº de proveedor:188602630
These sturdy, rigid racks feature interlocking end plates for secure stacking when loaded with tubes up to 10 cm (4") long.
- Polypropylene or polycarbonate construction
- Sturdy, rigid designs
- Decks may be white or clear depending on model
- Numbered and lettered rows
- Autoclavable
End plates have handles
The white surfaces of polypropylene racks make color comparisons easy. Sturdy and chemical-resistant, these racks will not shatter or stain. They are steam autoclavable at 121°C (250°F). Top shelf height is 7 cm (2³/₄").
Polycarbonate racks have crystal-clear decks and are perfect for submersing in water baths. Top shelf height is 7.3 cm (2⁷/₈"). Steam autoclavable at 121°C (250°F).
Rows are numbered and lettered for easy tube identification. Choose from polypropylene or polycarbonate to best suit your needs.