Large Capacity Laboratory Incubators, Reach-In, Shel Lab, Sheldon
Proveedor: SHELDON MANUFACTURING CO
Certificados
Acerca de este artículo
SHEL LAB large reach-in incubators provide over 30 cu.ft
chamber capacities while minimizing the amount of floor space used. Both models incorporate our microprocessor controller to achieve precise temperature uniformity. An independent, secondary temperature controller offers the added security of over temperature protection. The chamber floors are reinforced to support roller apparatus or shakers. Both models are supplied with six shelves capable of supporting the weight of benchtop instruments. One access port.
Información de pedido: Two-year limited warranty.
Especificaciones
- Catalog No:
- 89409-326
- 89409-324
- Intervalo de temperaturas:
- Ambient +8…70 °C
- Ambient +8…70 °C
- Uniformidad de la temperatura:
- +/-0.8°C at 37°C
- +/-0.8°C at 37°C
- No. of outlets:
- 4, internal
- 4, internal
- No. de charola:
- Six (6)
- Six (6)