- No. of outlets:Single One Amp
- Intervalo de temperaturas:Ambient +8…60 °C
- Capacidad:879 L (31 cu.ft.)
- Descripción:Large Capacity CO₂ Incubator
- Uniformidad de la temperatura:+/-0.5°C at 37°C
- Gama CO₂:0 - 20%
- Tiempo de recuperación:< 5 minutes (CO2 recovery rate)
- Referencia:89409-304
- Dimensiones interiores:83.1 x 66.0 x 160 cm (32.7 x 26.0 x 63")
- Dimensiones exteriores:100.4 x 85.8. x 191.2 cm (39.5 x 33.8 x 75.3")
- Detalles eléctricos:120 V, 50/60 Hz, 450 W, 4.5 A
- Nº de proveedor:SCO31
This units is well-suited for roller bottle apparatus and high-volume tissue culture applications and is ideal for cell harvesting.
- Accurracy: +/- 0.1% (CO₂ sensor)
This large capacity incubator maximizes laboratory space in a convenient floor model design. Its chamber floor is specifically designed for easy movement of roller bottle apparatus by use of a flip-out ramp. Supplied with a single one amp interior electrical outlet and gentle mechanical air convection that ensures excellent temperature uniformity, and eliminates cold spots. An infrared system accurately controls CO2 levels, provides fast CO2 recovery after door openings, and is not affected by temperature or humidity. This unit is supplied with six stainless steel shelves, which are adjustable on half inch increments.