Orders LinkContactUsLinkComponent
Sample Vials and Caps, MicroLiter, DWK Life Sciences
Sample Vials and Caps, MicroLiter, DWK Life Sciences
Sample Vials and Caps, MicroLiter, DWK Life Sciences
Certificates
Sample Vials and Caps, MicroLiter, DWK Life Sciences
Sample Vials and Caps, MicroLiter, DWK Life Sciences
2 Options Available Below
Product Details & Documents
All vials may appear to be created equal, but quality and performance make the difference when analyzing vital samples.
MicroLiter’s product line of sample vials offer superior storage and handling of larger volume samples. Available in clear and amber formats, with polypropylene or phenolic closures and a variety of septa and liner styles.
MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials. Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present. The result of this process are identical, clean, dependable vials, certified to ensure optimal performance and reproducible results.
Certifications: Certified Clean
Product Family Options
Vials
Información del producto
  • Capacity
    Diameter ×H
    Color
    Cap included
    Cap Size
    Availability
    Price
  • Vial
    4.00 ml
    15×45 mm
    Clear
    No
    13-425
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $816.00
    $816.00
    Paquete de 100
       $816.00
    Supplier Number: 13-1000
  • Vial
    4.00 ml
    15×45 mm
    Amber
    No
    13-425
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,318.63
    $1,318.63
    Paquete de 100
       $1,318.63
    Supplier Number: 13-2000
Caps
Información del producto
  • Catalog #
    Material
    Cap Size
    Septum
    Cap Color
    Cap Type
    Description
    Availability
    Price
  • PP
    13-425
    PTFE/Silicone
    Black
    Open top
    Caps
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,380.00
    $1,380.00
    Paquete de 100
       $1,380.00
Información del producto
  • Catalog #
    Septum
    Material
    Cap Color
    Cap Size
    Cap Type
    Description
    Availability
    Price
  • PP
    8-425
    Black
    Solid top
    Caps
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $219.00
    $219.00
    Paquete de 100
       $219.00