Orders LinkContactUsLinkComponent
Dial Hygrometer/Thermometer, Sper Scientific
Dial Hygrometer/Thermometer
  61220-785
 :  736920
Dial Hygrometer/Thermometer, Sper Scientific
  61220-785
Dial Hygrometer/Thermometer, Sper Scientific
Dial Hygrometer/Thermometer, Sper Scientific
Dial Hygrometer/Thermometer
$1,560.49
Each (1UN)
Lab or classroom hygrometer/thermometer.
  • Large, clear, 20–100% RH scale on 4" dial
  • Easily read from a distance
  • Wall mounts
  • Plastic face meets food industry requirements
The dial hygrometer/thermometer is ideal for classroom, lab or industrial use. Uses a precise synthetic hair hygrometer with an accuracy of ±5%. Inner dial shows temperature using a bimetal thermometer, with an accuracy of ±2°F. Weight 3.7 oz.
 : N.I.S.T. traceable certificate of compliance (for the certified model only).
  • Description:
    Dial Hygrometer/Thermometer