ContactUsLinkComponent
Detergents and Fertilizers as Pollutants: Studying an Algal Bloom Lab Activity
Detergents and Fertilizers as Pollutants: Studying an Algal Bloom Lab Activity
Catalog # 470190-664
Supplier:  Avantor
undefined
Detergents and Fertilizers as Pollutants: Studying an Algal Bloom Lab Activity
Catalog # 470190-664
Supplier:  Avantor
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Descripción:
    Detergents and Fertilizers as Pollutants: Studying an Algal Bloom Lab Activity
  • No. of Lab Groups:
    3
  • Referencia:
    470190-664
  • Nº de proveedor:
    WARD470190-664
Specifications
About this item
How Does Pollution Affect Our Waters?
  • Aligned to Fundamental Frameworks Concepts ESS3 and LS2
  • Pre and post laboratory assessments
  • Linked to a real world scenario
  • Estimated class/lab time required: 60 minutes over 2 lab sessions
This lab addresses the affects of phosphate and/or nitrate compounds on water. Students will observe water nitrate pollutant, phosphate pollutant over a given period of time. The students' understanding of this exercise will allow them to see the affects of pollution on water as well as how these specific pollutants affect the growth of algal cells. This activity utilizes Framework Science Standards ESS3 and LS2 (Earth and Human Activity and Ecosystems: Interactions, Energy, and Dynamics).
This kit has been aligned with all published National Standards. Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. This lab activity is designed for three groups of students. Time Requirement
Day 1: 30 Minutes; Day 10: 30 Minutes.
Ordering information: Materials include Nitrate pollutant Phosphorus pollutant, Plastic containers w/caps, Graduated pipets, Microscope slides, and Coverslips. Compound microscopes, Pond water, (or) Aged tap water and green algae culture are not included.