Orders LinkContactUsLinkComponent
Simulated ABO Blood Typing Lab Activity
Simulated ABO Blood Typing Lab Activity
Supplier:   Avantor
Simulated ABO Blood Typing Lab Activity
Supplier:  Avantor
Certificates
Simulated ABO Blood Typing Lab Activity
Simulated ABO Blood Typing Lab Activity
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ever Wonder if your Blood is Type A, Type B, or Something Else?
  • Learn Blood Typing Without Using Real Blood
  • NGSS Life Science DCIs LS1 and LS3
  • Pre And Post-Laboratory Assessments
  • Links To Real World Concepts
  • Time Requirements: Part A: 30 minutes, Part B: 20 minutes
In this lab activity, students explore the core life science concepts of Structures and Properties (LS1) and Heredity (LS3) while determining the blood type of WARD'S completely safe and non-toxic Simulated Blood. Standard blood typing procedures are used to teach students about the different blood types as well as antigen/antibody relationships, and how these relationships effect the results of the blood typing procedure. This kit has been aligned with all published National Standards. Pre- and Post-laboratory assessments and vocabulary words all target specific Science and Engineering Practices and common core standards. The teacher guide also contains specific "Did You Know" concepts that link the real world with the scientific standards explored. A refill kit is available and contains only consumable component items it does not contain additional student or teacher guides.
Accessories information: Refill kit includes simulated blood samples and antisera.
Ordering information: Kit contents include: WARD'S Simulated Blood Type A, WARD'S Simulated Blood Type B, WARD'S Simulated Blood Type AB, WARD'S Simulated Blood Type O, Simulated Anti-A Serum, Simulated Anti-B Serum, Blood Typing Slides, Toothpicks, Teachers guide, and Student copy masters. Materials needed but not included: Compound Microscopes (400X magnification), Microscope Slide, Coverslips, and Markers.
Product Family Options
Información del producto
  • No. of Lab Groups
    Availability
    Price
  • 470190-616
    Simulated ABO Blood Typing Kit
    Simulated ABO Blood Typing Kit
    12
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $1,162.65
    $1,162.65
    Kit de 1
       $1,162.65
  • 470015-186
    Simulated ABO Blood Typing Kit, Refill
    Simulated ABO Blood Typing Kit, Refill
    12
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $634.80
    $634.80
    Kit de 1
       $634.80
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.