PIG® Maintenance Wipers, New Pig
89069-050
- Color:Blue
- Referencia:89069-050
- Empaquetado:1/4-fold
- An x La:30.5 x 33 cm (12 x 13")
- Nº de proveedor:WIP218
Indispensable in machine shops, fleet garages and plants; unaffected by brake fluid, antifreeze, grease, oils and other liquids.
- Low-lint material won't leave residue or create a mess
- Lower cost per wiper
- Excellent for low-cost daily maintenance (not recommended for long-term contact with solvents)
Embossed surface grabs grime for tough cleaning with backup from 2-ply tissue and hidden nylon scrim.
Available ¹/₄ folded and poly wrapped for easy handling and loading. Convenient pop-up box lets you take just one wiper at a time while keeping remaining stock clean and dry.