ContactUsLinkComponent
Sugar Refractometer, 0 to 80%, Sper Scientific
Sugar Refractometer, 0 to 80%, Sper Scientific
Catalog # 66214-980
Supplier:  SPER SCIENTIFIC
undefined
Sugar Refractometer, 0 to 80%, Sper Scientific
Catalog # 66214-980
Supplier:  SPER SCIENTIFIC
Supplier Number:  300003
Specifications
  • Descripción:
    Sugar Refractometer 0–80%
  • Referencia:
    66214-980
  • Nº de proveedor:
    300003
Specifications
About this item
This unit covers all of the substances of the Sugar-Brix Refractometer 0–32% and Sugar Refractometer, 28–62%, as well as very high sugar content products such as jam, marmalade, and syrup
Its wide range also makes this refractometer a good choice for the testing of any solution with an unknown sugar content. To operate the device, simply place a drop or two of sample on the prism and read the results.
Ordering information: Refractometer is supplied with a hard carrying case, calibration tool, and 5-year warranty.