Stand and Rod Deluxe Pack
Catalog # 470137-894
Supplier: EISCO SCIENTIFIC LLC
Specifications
- DescripciĂłn:Stand and Rod Deluxe Pack
- Referencia:470137-894
- NÂș de proveedor:STDRD1
Specifications
About this item
Solid and Sturdy
The base in this pack is base cut steel and is a heavier choice than the standard version. The powder-coated base has rubber backing to prevent slipping. Rod diameter is 12mm and they thread directly into the base.